accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3NNHNONCl

CAS RN 4443-99-6

IUPAC Name: 3-(diethylamino)-7-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazin-1-yl]-5-phenyl-5λ⁵-phenazin-5-ylium chloride
Synonyms: basic black 2
Molecular Weight (g/mol): 484
Molecular Formula: C28H26ClN5O
InChi Key: VJDDAARZIFHSQY-UHFFFAOYSA-N
SMILES: [Cl-].CCN(CC)C1=CC=C2N=C3C=CC(NN=C4C=CC(=O)C=C4)=CC3=[N+](C3=CC=CC=C3)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results