accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4450-98-0

IUPAC Name: ethyl 1-benzyl-4-hydroxy-5-oxo-2,5-dihydro-1H-pyrrole-3-carboxylate
Synonyms: ethyl 1-benzyl-4-hydroxy-5-oxo-2H-pyrrole-3-carboxylate
Molecular Weight (g/mol): 261.28
Molecular Formula: C14H15NO4
InChi Key: IGYRPDIWSYGHMY-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(O)C(=O)N(CC2=CC=CC=C2)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results