accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4454-05-1

IUPAC Name: 2-methoxy-3,4-dihydro-2H-pyran
Synonyms: 2-methoxy-3,4-dihydro-2H-pyran
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: XCYWUZHUTJDTGS-UHFFFAOYNA-N
SMILES: COC1CCC=CO1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)