Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dihydro-2-methoxy-2H-pyran, +99%, Thermo Scientific™

Catalog No. AC177621000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC177621000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC177621000 Supplier Thermo Scientific Chemicals Supplier No. 177621000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4454-05-1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0000g/mL
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C6H10O2
Refractive Index 1.4410 to 1.4430 (20°C, 589nm)
Quantity 100 g
MDL Number MFCD00006560
Synonym 3,4-dihydro-2-methoxy-2h-pyran, 2h-pyran, 3,4-dihydro-2-methoxy, 2,3-dihydro-2-methoxy 4h pyran, 2-methoxy-2,3-dihydro-4h-pyran, acmc-1ahgh, dsstox_cid_7570, dsstox_rid_78508, dsstox_gsid_27570, ksc496o2j
InChI Key XCYWUZHUTJDTGS-UHFFFAOYNA-N
SMILES COC1CCC=CO1
IUPAC Name 2-methoxy-3,4-dihydro-2H-pyran
Molecular Weight (g/mol) 114.14
PubChem CID 91538
Formula Weight 114.14
Percent Purity ≥99.0%
Physical Form Liquid
Chemical Name or Material 3, 4-Dihydro-2-methoxy-2H-pyran, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Toxic if inhaled.
Harmful if swallowed.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-698-3
RTECSNumber UP7900000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.