accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNHONHFFFOHNONHHNOSCH3

CAS RN 445479-97-0

IUPAC Name: 4-(methylsulfanyl)-N-(2-{2-[(2-{[(propan-2-yl)carbamoyl]amino}-5-(trifluoromethyl)phenyl)formamido]acetamido}cyclohexyl)benzamide
Synonyms: N-{2-[2-({2-[(isopropylcarbamoyl)amino]-5-(trifluoromethyl)phenyl}formamido)acetamido]cyclohexyl}-4-(methylsulfanyl)benzamide
Molecular Weight (g/mol): 593.67
Molecular Formula: C28H34F3N5O4S
InChi Key: IBPXYDUJQWENPM-UHFFFAOYNA-N
SMILES: CSC1=CC=C(C=C1)C(=O)NC1CCCCC1NC(=O)CNC(=O)C1=C(NC(=O)NC(C)C)C=CC(=C1)C(F)(F)F
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)