accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4462-96-8

IUPAC Name: 3-oxabicyclo[3.2.0]heptane-2,4-dione
Synonyms: 3-oxabicyclo[3.2.0]heptane-2,4-dione
Molecular Weight (g/mol): 126.11
Molecular Formula: C6H6O3
InChi Key: NMNZZIMBGSGRPN-UHFFFAOYNA-N
SMILES: O=C1OC(=O)C2CCC12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results