Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perhydrocyclobuta[c]furan-1,3-dione, 97%, Thermo Scientific™

Catalog No. BTB13668DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB13668DE 5 g
BTB13668CB 250 mg
BTB13668DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BTB13668DE Supplier Thermo Scientific Chemicals Supplier No. BTB13668DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4462-96-8
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.111
InChI Key NMNZZIMBGSGRPN-UHFFFAOYSA-N
Synonym 3-oxabicyclo 3.2.0 heptane-2,4-dione, perhydrocyclobuta c furan-1,3-dione, 1,2-cyclobutanedicarboxylic anhydride, cyclobutane-1,2-dicarboxylic anhydride, 1,2-cyclobutanedicarboxylic anhydride, cis-,, acmc-1ahmk, 3-oxabicyclo 3.2.0 heptane-2, cis
PubChem CID 138261
IUPAC Name 3-oxabicyclo[3.2.0]heptane-2,4-dione
SMILES C1CC2C1C(=O)OC2=O

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material Perhydrocyclobuta[c]furan-1, 3-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.