CAS RN 4472-41-7
N,N-Dimethylformamide-d7, for NMR, packaged in 0.75 ml ampoules, 99.5 atom % D
CAS: 4472-41-7 Molecular Formula: C3H7NO Molecular Weight (g/mol): 80.138 MDL Number: MFCD00003286 InChI Key: ZMXDDKWLCZADIW-YYWVXINBSA-N Synonym: n,n-dimethylformamide-d7,dmf-d7,deuterated dmf,n,n-di 2h3 methyl 2h formamide,dimethyl formamide-d7,dimethylformamide d,heptadeutero-n,n-dimethylformamide,n,n-dimethylformamide-d7 dmf-d7,formamide-1-d, n,n-di methyl-d3,n,n-dimethylformamide-d7 99.5atom%d PubChem CID: 78225 IUPAC Name: 1-deuterio-N,N-bis(trideuteriomethyl)formamide SMILES: CN(C)C=O
N,N-Dimethylformamide-d7, for NMR, 99.5 atom % D, Thermo Scientific Chemicals
CAS: 4472-41-7 Molecular Formula: C3H7NO MDL Number: MFCD00003286 InChI Key: ZMXDDKWLCZADIW-YYWVXINBSA-N Synonym: dimethylformamide d,formamide-1-d, n,n-di methyl-d3,deuterated dmf,n,n-dimethylformamide-d7,n,n-dimethylformamide-d7 99.5atom%d,n,n-dimethylformamide-d7 dmf-d7,dimethyl formamide-d7,heptadeutero-n,n-dimethylformamide,dmf-d7,n,n-di 2h3 methyl 2h formamide PubChem CID: 78225
N,N-Dimethylformamide-d{7}, 99.5% (Isotopic)
CAS: 4472-41-7 Molecular Formula: C3H7NO Molecular Weight (g/mol): 80.138 MDL Number: MFCD00003286 InChI Key: ZMXDDKWLCZADIW-YYWVXINBSA-N Synonym: n,n-dimethylformamide-d7,dmf-d7,deuterated dmf,n,n-di 2h3 methyl 2h formamide,dimethyl formamide-d7,dimethylformamide d,heptadeutero-n,n-dimethylformamide,n,n-dimethylformamide-d7 dmf-d7,formamide-1-d, n,n-di methyl-d3,n,n-dimethylformamide-d7 99.5atom%d PubChem CID: 78225 IUPAC Name: 1-deuterio-N,N-bis(trideuteriomethyl)formamide SMILES: CN(C)C=O
Dimethylformamide - D7, deuteration degree min. 99.5% for NMR spectroscopy MagniSolv™, MilliporeSigma™
CAS: 4472-41-7 Molecular Formula: C3H7NO Molecular Weight (g/mol): 80.138 InChI Key: ZMXDDKWLCZADIW-YYWVXINBSA-N Synonym: n,n-dimethylformamide-d7,dmf-d7,deuterated dmf,n,n-di 2h3 methyl 2h formamide,dimethyl formamide-d7,dimethylformamide d,heptadeutero-n,n-dimethylformamide,n,n-dimethylformamide-d7 dmf-d7,formamide-1-d, n,n-di methyl-d3,n,n-dimethylformamide-d7 99.5atom%d PubChem CID: 78225 IUPAC Name: 1-deuterio-N,N-bis(trideuteriomethyl)formamide SMILES: CN(C)C=O