Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide-d7, for NMR, 99.5 atom % D

Catalog No. AC183600250
Change view
Click to view available options
Quantity:
1 mL
5 mL
10 mL
25 mL
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC183600250 25 mL Glass Bottle
AC183600010 1 mL Glass Bottle
AC183600050 5 mL Glass Bottle
AC183600100 10 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AC183600250 Supplier Thermo Scientific Chemicals Supplier No. 183600250
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4472-41-7 | C3H7NO | 80.138 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4472-41-7
Molecular Formula C3H7NO
MDL Number MFCD00003286
InChI Key ZMXDDKWLCZADIW-YYWVXINBSA-N
Synonym dimethylformamide d, formamide-1-d, n,n-di methyl-d3, deuterated dmf, n,n-dimethylformamide-d7, n,n-dimethylformamide-d7 99.5atom%d, n,n-dimethylformamide-d7 dmf-d7, dimethyl formamide-d7, heptadeutero-n,n-dimethylformamide, dmf-d7, n,n-di 2h3 methyl 2h formamide
PubChem CID 78225

Specifications

Physical Form Liquid
Color Colorless
Packaging Glass Bottle
Deuterium (D) 99.4% min.
Flash Point 58°C
Quantity 25 mL
Density 1.0300 g/mL
Refractive Index 1.428
Water 0.1% max. (Coulometric)
Linear Formula DCON(CD3)2
SMILES [2H]C(=O)N(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC Name N,N-di(²H₃)methyl(²H)formamide
Molecular Weight (g/mol) 80.14
Formula Weight 80.15
Grade NMR
Chemical Name or Material N, N-Dimethylformamide-d7, 99.5 atom % D
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:

Harmful if swallowed.

Causes serious eye irritation.

Harmful in contact with skin.

Harmful if inhaled.

May damage the unborn child.

Flammable liquid and vapor.
Precautionary Statement GHS P Statement:

Wear protective gloves/protective clothing/eye protection/face protection.

Obtain special instructions before use.

IF exposed or concerned: Get medical advice/attention.

IF ON SKIN: Wash with plenty of soap and water.

IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Keep away from heat/sparks/open flames/hot surfaces. - No smoking.



WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-745-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.