Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylformamide - D7, deuteration degree min. 99.5% for NMR spectroscopy MagniSolv™, MilliporeSigma™

Catalog No. M1116560009 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
10 x 0.75 mL
Packaging:
Glass Ampule
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1116560009 10 x 0.75 mL Glass Ampule
M1116560001 1 mL Glass Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1116560009 Supplier MilliporeSigma Supplier No. 1.11656.0009
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For NMR Spectroscopy

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4472-41-7
Molecular Formula C3H7NO
Molecular Weight (g/mol) 80.138
InChI Key ZMXDDKWLCZADIW-YYWVXINBSA-N
Synonym n,n-dimethylformamide-d7, dmf-d7, deuterated dmf, n,n-di 2h3 methyl 2h formamide, dimethyl formamide-d7, dimethylformamide d, heptadeutero-n,n-dimethylformamide, n,n-dimethylformamide-d7 dmf-d7, formamide-1-d, n,n-di methyl-d3, n,n-dimethylformamide-d7 99.5atom%d
PubChem CID 78225
IUPAC Name 1-deuterio-N,N-bis(trideuteriomethyl)formamide
SMILES CN(C)C=O

Specifications

Physical Form Liquid
Color Colorless
Packaging Glass Ampule
Odor Weakly Amine-like
Quantity 10 x 0.75 mL
Vapor Pressure 3.77 hPa at 20°C (68°F)
Assay Percent Range 99.5% min.
Formula Weight 80.14
Chemical Name or Material Dimethylformamide - D7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.