accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NH2

CAS RN 44745-29-1

IUPAC Name: heptan-2-amine
Synonyms: tuaminoheptane tuaminoheptano heptamine (sympathomimetic) tuaminoheptanum armeen L-7 tuaminheptanum tuaminoheptane (inn), its isomers and salts tuaminoheptane (substance)
Molecular Weight (g/mol): 115.22
Molecular Formula: C7H17N
InChi Key: VSRBKQFNFZQRBM-UHFFFAOYNA-N
SMILES: CCCCCC(C)N
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results