accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFHNOHNFFFOHOH

CAS RN 448895-37-2

IUPAC Name: 1,3-bis[2-hydroxy-5-(trifluoromethyl)phenyl]urea
Synonyms: 1,3-bis[2-hydroxy-5-(trifluoromethyl)phenyl]urea
Molecular Weight (g/mol): 380.25
Molecular Formula: C15H10F6N2O3
InChi Key: NJFVQMRYJZHGME-UHFFFAOYSA-N
SMILES: OC1=C(NC(=O)NC2=C(O)C=CC(=C2)C(F)(F)F)C=C(C=C1)C(F)(F)F
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results