accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOSNOHN

CAS RN 448906-42-1

IUPAC Name: methyl 2-(1H-indole-3-carbonyl)-1,3-thiazole-4-carboxylate
Synonyms: methyl 2-(1H-indole-3-carbonyl)-1,3-thiazole-4-carboxylate
Molecular Weight (g/mol): 286.31
Molecular Formula: C14H10N2O3S
InChi Key: KDDXOGDIPZSCTM-UHFFFAOYSA-N
SMILES: COC(=O)C1=CSC(=N1)C(=O)C1=CNC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)