accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CNCH3H3CNHOCH2

CAS RN 45021-77-0

IUPAC Name: trimethyl[3-(prop-2-enamido)propyl]azanium chloride
Synonyms: trimethyl[3-(prop-2-enamido)propyl]azanium chloride
Molecular Weight (g/mol): 206.71
Molecular Formula: C9H19ClN2O
InChi Key: OEIXGLMQZVLOQX-UHFFFAOYSA-N
SMILES: [Cl-].C[N+](C)(C)CCCNC(=O)C=C
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)