Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-Acrylamidopropyl)trimethylammonium Chloride (74-76% in Water) (stabilized with MEHQ), TCI America™
SDP

Catalog No. A149325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A149325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A149325ML Supplier TCI America Supplier No. A149325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 45021-77-0
Molecular Formula C9H19ClN2O
Molecular Weight (g/mol) 206.714
MDL Number MFCD00274249
InChI Key OEIXGLMQZVLOQX-UHFFFAOYSA-N
PubChem CID 170750
IUPAC Name trimethyl-[3-(prop-2-enoylamino)propyl]azanium;chloride
SMILES C[N+](C)(C)CCCNC(=O)C=C.[Cl-]

Specifications

Quantity 25 mL
Formula Weight 206.71
Physical Form Liquid
Chemical Name or Material (3-Acrylamidopropyl)trimethylammonium Chloride (74-76% in Water) (stabilized with MEHQ)
EINECSNumber (2)-3493
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.