accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2COOFFFF

CAS RN 45102-52-1

IUPAC Name: 2,2,3,3-tetrafluoropropyl 2-methylprop-2-enoate
Synonyms: 2,2,3,3-tetrafluoropropyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 200.13
Molecular Formula: C7H8F4O2
InChi Key: RSVZYSKAPMBSMY-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCC(F)(F)C(F)F
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results