accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH3COCH3ClO

CAS RN 4521-61-3

IUPAC Name: 3,4,5-trimethoxybenzoyl chloride
Synonyms: 3,4,5-trimethoxybenzoyl chloride
Molecular Weight (g/mol): 230.64
Molecular Formula: C10H11ClO4
InChi Key: BUHYMJLFRZAFBF-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(OC)=C1OC)C(Cl)=O
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)