Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Trimethoxybenzoyl Chloride 97.0+%, TCI America™
SDP

Catalog No. T184225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T184225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T184225G Supplier TCI America Supplier No. T184225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4521-61-3
Molecular Formula C10H11ClO4
Molecular Weight (g/mol) 230.644
MDL Number MFCD00000675
InChI Key BUHYMJLFRZAFBF-UHFFFAOYSA-N
Synonym benzoyl chloride, 3,4,5-trimethoxy, trimethylgalloyl chloride, tri-o-methylgalloyl chloride, 3,4,5-trimethoxybenzoylchloride, zlchem 339, pubchem22153, acmc-1ahlk, 3,5-trimethoxybenzoyl chloride, timtec-bb sbb040629, 3,4,5-trimethoxy-benzoylchloride
PubChem CID 78282
IUPAC Name 3,4,5-trimethoxybenzoyl chloride
SMILES COC1=CC(=CC(=C1OC)OC)C(=O)Cl

Specifications

Melting Point 78°C
Color White-Yellow
Boiling Point 185°C
Quantity 25 g
UN Number 3261
Formula Weight 230.64
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5-Trimethoxybenzoyl Chloride
EINECSNumber (3)-1683
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.