accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOFNOO

CAS RN 453-72-5

IUPAC Name: 1-fluoro-4-methanesulfonyl-2-nitrobenzene
Synonyms: 1-fluoro-4-methanesulfonyl-2-nitrobenzene
Molecular Weight (g/mol): 219.19
Molecular Formula: C7H6FNO4S
InChi Key: OUSNDSFSTBZESM-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=C(F)C(=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)