Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-5-methylsulphonylnitrobenzene, 95%, Thermo Scientific™

Catalog No. AC425000025
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC425000025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC425000025 Supplier Thermo Scientific Chemicals Supplier No. 425000025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 453-72-5
Molecular Formula C7H6FNO4S
Molecular Weight (g/mol) 219.19
MDL Number MFCD00025066
InChI Key OUSNDSFSTBZESM-UHFFFAOYSA-N
Synonym 4-methylsulfonyl-2-nitrofluorobenzene, 1-fluoro-4-methylsulfonyl-2-nitrobenzene, 1-fluoro-4-methanesulfonyl-2-nitrobenzene, 2-fluoro-5-methyl sulfonyl nitrobenzene, 2-fluoro-5-methylsulphonylnitrobenzene, 4-fluoro-3-nitrophenyl methyl sulfone, benzene,1-fluoro-4-methylsulfonyl-2-nitro, 2-fluoro-5-methylsulfonylnitrobenzene, 2-fluoro-5-methylsulphonyl nitrobenzene, 1-fluoro-4-methylsulfonyl-2-nitro-benzene
PubChem CID 313137
IUPAC Name 1-fluoro-4-methanesulfonyl-2-nitrobenzene
SMILES CS(=O)(=O)C1=CC=C(F)C(=C1)[N+]([O-])=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 152°C to 158°C
Color Green-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 2.5 g
Formula Weight 219.19
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-5-methylsulphonylnitrobenzene, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.