accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CNHCH3Cl

CAS RN 4535-88-0

IUPAC Name: (2-chloroethyl)(ethyl)methylazanium chloride
Synonyms: (2-chloroethyl)(ethyl)methylazanium chloride
Molecular Weight (g/mol): 158.07
Molecular Formula: C5H13Cl2N
InChi Key: JNZADWSFSFZFLJ-UHFFFAOYNA-N
SMILES: [Cl-].CC[NH+](C)CCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results