Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2(Methylethylamino)ethyl Chloride HCl, MP Biomedicals

Catalog No. ICN226334 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN226334 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN226334 Supplier MP Biomedicals, Inc Supplier No. 0522633410

Chemical Identifiers

CAS 4535-88-0
Molecular Formula C5H13Cl2N
Molecular Weight (g/mol) 158.07
InChI Key JNZADWSFSFZFLJ-UHFFFAOYNA-N
Synonym jnzadwsfsfzflj-uhfffaoysa-n, 2-chloroethyl-ethyl-methylazanium,chloride, 2 methylethylamino ethyl chloride hcl, 2-chloro-n,n-diethyl-methylamine hydrochloride, 2 methylethylamino ethyl chloride hydrochloride, 2-n-methyl-n-ethylamino ethyl chloride hydrochloride
PubChem CID 25021949

Specifications

Quantity 10 g
Formula Weight 158.07
Chemical Name or Material 2(Methylethylamino)Ethyl Chloride HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.