accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4542-57-8

IUPAC Name: 1-(dodecyloxy)dodecane
Synonyms: dilauryl ether
Molecular Weight (g/mol): 354.66
Molecular Formula: C24H50O
InChi Key: CMCBDXRRFKYBDG-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCOCCCCCCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results