Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecyl Ether 95.0+%, TCI America™
SDP

Catalog No. D099425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D099425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D099425G Supplier TCI America Supplier No. D099425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4542-57-8
Molecular Formula C24H50O
Molecular Weight (g/mol) 354.66
MDL Number MFCD00026590
InChI Key CMCBDXRRFKYBDG-UHFFFAOYSA-N
Synonym Didodecyl Ether, Dilauryl Ether, Lauryl Ether
PubChem CID 20667
IUPAC Name 1-(dodecyloxy)dodecane
SMILES CCCCCCCCCCCCOCCCCCCCCCCCC

Specifications

Melting Point 32°C
Color White
Boiling Point 190°C
Quantity 25 g
Formula Weight 354.66
Percent Purity ≥95.0% (GC)
Physical Form Wax
Chemical Name or Material Dodecyl Ether
EINECSNumber (2)-3076
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.