accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4543-33-3

IUPAC Name: 1,3,6,8-tetranitro-9H-carbazole
Synonyms: 1,3,6,8-tetranitro-9H-carbazole
Molecular Weight (g/mol): 347.2
Molecular Formula: C12H5N5O8
InChi Key: JUSWGNJYSBSOFM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC2=C(NC3=C2C=C(C=C3[N+]([O-])=O)[N+]([O-])=O)C(=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results