accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHOH

CAS RN 4543-95-7

IUPAC Name: 4-(butylamino)butan-1-ol
Synonyms: 4-(butylamino)butan-1-ol
Molecular Weight (g/mol): 145.25
Molecular Formula: C8H19NO
InChi Key: OPUJAKVDYGQVHP-UHFFFAOYSA-N
SMILES: CCCCNCCCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results