accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 454482-11-2

BOOH3CCH3H3CH3CNCH3

CAS RN 454482-11-2

IUPAC Name: 1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine
Synonyms: 1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine
Molecular Weight (g/mol): 223.12
Molecular Formula: C12H22BNO2
InChi Key: SQMVRFXDBRYXFQ-UHFFFAOYSA-N
SMILES: CN1CCC(=CC1)B1OC(C)(C)C(C)(C)O1
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results