accessibility menu, dialog, popup

UserName

Viewing profile as user:
FO

CAS RN 455-32-3

IUPAC Name: benzoyl fluoride
Synonyms: benzoyl fluoride
Molecular Weight (g/mol): 124.11
Molecular Formula: C7H5FO
InChi Key: HPMLGNIUXVXALD-UHFFFAOYSA-N
SMILES: FC(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)