Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoyl fluoride, 97%

Catalog No. AAL1146414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1146414 25 g
AAL1146406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1146414 Supplier Thermo Scientific Chemicals Supplier No. L1146414
  • $209.00
    $167.65 / Each of 1

    Save $41.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 455-32-3 | C7H5FO | 124.11 g/mol

Benzoyl fluoride is used as a mild fluorinating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzoyl fluoride is used as a mild fluorinating agent.

Solubility
Hydrolyses in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to moisture. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 455-32-3
Molecular Formula C7H5FO
Molecular Weight (g/mol) 124.11
MDL Number MFCD00000364
InChI Key HPMLGNIUXVXALD-UHFFFAOYSA-N
Synonym benzoylfluoride, unii-lor25i34hd, lor25i34hd, benzoylfluorid, acmc-1alcy, hpmlgniuxvxald-uhfffaoysa
PubChem CID 67999
IUPAC Name benzoyl fluoride
SMILES FC(=O)C1=CC=CC=C1

Specifications

Melting Point ∼-28°C
Density 1.14
Boiling Point 159°C to 161°C
Flash Point 48°C (118°F)
Odor Pungent
Refractive Index 1.496
Quantity 25 g
UN Number UN2920
Beilstein 1904977
Sensitivity Moisture sensitive
Solubility Information Hydrolyses in water.
Formula Weight 124.12
Percent Purity 98%
Chemical Name or Material Benzoyl fluoride
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 207-244-9
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.