Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoyl fluoride, 97%

Catalog No. AC402041000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC402041000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC402041000 Supplier Thermo Scientific Chemicals Supplier No. 402041000

missing translation for 'validMainframeMsg'

CAS: 455-32-3 | C7H5FO | 124.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 455-32-3
Molecular Formula C7H5FO
Molecular Weight (g/mol) 124.11
MDL Number MFCD00000364
InChI Key HPMLGNIUXVXALD-UHFFFAOYSA-N
Synonym benzoylfluoride, unii-lor25i34hd, lor25i34hd, benzoylfluorid, acmc-1alcy, hpmlgniuxvxald-uhfffaoysa
PubChem CID 67999
IUPAC Name benzoyl fluoride
SMILES FC(=O)C1=CC=CC=C1

Specifications

Melting Point -28.0°C
Density 1.1500g/mL
Boiling Point 159.0°C to 161.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Refractive Index 1.496
Linear Formula C6H5COF
Quantity 100 g
Specific Gravity 1.15
Solubility Information Solubility in water: hydrolyses.
Formula Weight 124.11
Percent Purity ≥96.0% (GC)
Chemical Name or Material Benzoyl fluoride
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement Flammable liquid and vapor. Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.