accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4569-45-3

IUPAC Name: 9,9-dimethyl-9H-fluorene
Synonyms: 9,9-dimethylfluorene
Molecular Weight (g/mol): 194.28
Molecular Formula: C15H14
InChi Key: ZHQNDEHZACHHTA-UHFFFAOYSA-N
SMILES: CC1(C)C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)