Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Dimethylfluorene (purified by sublimation) 99.0+%, TCI America™
SDP

Catalog No. D38101G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D38101G Supplier TCI America Supplier No. D38101G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4569-45-3
Molecular Formula C15H14
Molecular Weight (g/mol) 194.28
MDL Number MFCD00114670,MFCD31699973
InChI Key ZHQNDEHZACHHTA-UHFFFAOYSA-N
PubChem CID 78325
IUPAC Name 9,9-dimethyl-9H-fluorene
SMILES CC1(C)C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 97°C
Color White
Boiling Point 287°C
Quantity 1 g
Formula Weight 194.28
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Dimethylfluorene (purified by sublimation)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.