accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 457-68-1

FF

CAS RN 457-68-1

IUPAC Name: 1-fluoro-4-[(4-fluorophenyl)methyl]benzene
Synonyms: 1-fluoro-4-[(4-fluorophenyl)methyl]benzene
Molecular Weight (g/mol): 204.22
Molecular Formula: C13H10F2
InChi Key: DXQVFHQUHOFROC-UHFFFAOYSA-N
SMILES: FC1=CC=C(CC2=CC=C(F)C=C2)C=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)