Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Difluorodiphenylmethane 99.0+%, TCI America™
SDP

Catalog No. D192525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D192525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D192525G Supplier TCI America Supplier No. D192525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 457-68-1
Molecular Formula C13H10F2
Molecular Weight (g/mol) 204.22
MDL Number MFCD00000360
InChI Key DXQVFHQUHOFROC-UHFFFAOYSA-N
Synonym bis 4-fluorophenyl methane, 4,4'-difluorodiphenylmethane, 1,1'-methylenebis 4-fluorobenzene, 1-fluoro-4-4-fluorophenyl methyl benzene, benzene, 1,1'-methylenebis 4-fluoro, 4-fluoro-1-4-fluorophenyl methyl benzene, 4,4'-difluorodiphenylamine, bis-4-fluorophenyl methane, 4,4'-difluorodiphenyl methane, 1,1-methylenebis 4-fluorobenzene
PubChem CID 96094
IUPAC Name 1-fluoro-4-[(4-fluorophenyl)methyl]benzene
SMILES C1=CC(=CC=C1CC2=CC=C(C=C2)F)F

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 204.22
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4,4′-Difluorodiphenylmethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.