accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaNH2NNNN(R)O(R)OPOOO(S)HO(R)HO

CAS RN 4578-31-8

IUPAC Name: disodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
Synonyms: disodium adenosine phosphate
Molecular Weight (g/mol): 391.19
Molecular Formula: C10H12N5Na2O7P
InChi Key: QGXLVXZRPRRCRP-IDIVVRGQSA-L
SMILES: [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
Quantity 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)