Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine-5'-Monophosphate Disodium Salt Hydrate, ≥99%, MP Biomedicals™

Catalog No. ICN15026425 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15026425 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15026425 Supplier MP Biomedicals, Inc Supplier No. 0215026425
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Source: yeast

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4578-31-8
Molecular Formula C10H12N5Na2O7P
Molecular Weight (g/mol) 391.19
MDL Number MFCD00065023
InChI Key QGXLVXZRPRRCRP-IDIVVRGQSA-L
Synonym adenosine 5'-monophosphate disodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate, disodium amp, amp disodium salt, disodium 5'-amp, adenosine phosphate disodium, 5'-amp disodium salt, 5'-adenylic acid, disodium salt, unii-t1wz11dsrn, adenylic acid disodium salt
PubChem CID 20712
ChEBI CHEBI:81690
IUPAC Name disodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
SMILES [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Color White
Assay Percent Range ≥99%
Quantity 25 g
Formula Weight 391.2
Percent Purity ≥99%
Grade Not Available
Physical Form Powder
Chemical Name or Material Adenosine-5'-Monophosphate Disodium Salt Hydrate
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.