accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 459-56-3

IUPAC Name: (4-fluorophenyl)methanol
Synonyms: benzenemethanol, 4-fluoro-
Molecular Weight (g/mol): 126.13
Molecular Formula: C7H7FO
InChi Key: GEZMEIHVFSWOCA-UHFFFAOYSA-N
SMILES: OCC1=CC=C(F)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)