Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorobenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. F02515G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02515G 5 g
F025125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02515G Supplier TCI America Supplier No. F02515G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 459-56-3
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00004651
InChI Key GEZMEIHVFSWOCA-UHFFFAOYSA-N
Synonym 4-fluorobenzyl alcohol, 4-fluorophenyl methanol, benzenemethanol, 4-fluoro, p-fluorobenzyl alcohol, 4-fluorobenzylalcohol, 4-fluorobenzylic alcohol, ccris 5115, 4-fluorobenzoic alcohol, 4-fluoro benzyl alcohol, 4-fluorophenyl methan-1-ol
PubChem CID 68022
IUPAC Name (4-fluorophenyl)methanol
SMILES C1=CC(=CC=C1CO)F

Specifications

Color White-Yellow
Boiling Point 99°C
Quantity 5 g
Formula Weight 126.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Fluorobenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.