CAS RN 459841-96-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
R&D Systems™ ONO 8130
A Tocris product, part of R&D Systems. EP1 receptor antagonist