accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNH

CAS RN 46040-83-9

IUPAC Name: 3,3'-bipiperidine
Synonyms: 3,3'-bipiperidine
Molecular Weight (g/mol): 168.28
Molecular Formula: C10H20N2
InChi Key: IKNFMHZKPYNCLN-UHFFFAOYNA-N
SMILES: C1CNCC(C1)C1CCCNC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results