accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNSOOH3COCH3OCH3

CAS RN 461000-66-8

IUPAC Name: 3,4,5-trimethoxy-N-(2-methoxyethyl)-N-(4-phenyl-1,3-thiazol-2-yl)benzamide
Synonyms: 3,4,5-trimethoxy-N-(2-methoxyethyl)-N-(4-phenyl-1,3-thiazol-2-yl)benzamide
Molecular Weight (g/mol): 428.5
Molecular Formula: C22H24N2O5S
InChi Key: ZUXNHFFVQWADJL-UHFFFAOYSA-N
SMILES: COCCN(C(=O)C1=CC(OC)=C(OC)C(OC)=C1)C1=NC(=CS1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)