accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4637-24-5

IUPAC Name: (dimethoxymethyl)dimethylamine
Synonyms: (dimethoxymethyl)dimethylamine
Molecular Weight (g/mol): 119.16
Molecular Formula: C5H13NO2
InChi Key: ZSXGLVDWWRXATF-UHFFFAOYSA-N
SMILES: COC(OC)N(C)C
Boiling Point 
  • (4)
  • (3)
  • (4)
  • (4)
Molecular Weight (g/mol) 
  • (11)
  • (4)
Quantity 
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (1)
  • (3)
  • (4)
Physical Form 
  • (7)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (4)
  • (4)
  • (7)
  • (4)
  • (3)
  • (4)
  • (4)
  • (11)
  • (4)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (4)
  • (7)
  • (1)