Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N-Dimethylformamide Dimethyl Acetal

Catalog No. 140732500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
2.5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-073-2500ML 500 mL
14-073-225L 2.5 L
14-073-225ML 25 mL
14-073-2100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 14-073-2500ML Supplier Sigma Aldrich Supplier No. 140732500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N,N-Dimethylformamide dimethyl acetal (DMF-DMA) can be used as: An organocatalyst for the synthesis of 1,2-disubstituted-3,4-dihydronaphthalenes by the reaction of vinylarenes and electron-deficient alkynes A building block for the preparation of heterocyclic compounds such as enamines and amidines via formylation of methylene and amino groups A reagent in the synthesis of 1,4-diaryl-piperazine-2,5-diones by reacting with chloroacetamides A reagent along with enamine and alkyne for the preparation of 2,3,4,5-tetrasubstituted pyridine derivatives by three-component coupling reaction in the presence of Me3SiCl.

Specifications

CAS 4637-24-5
Density 0.897 g/mL at 25°C
Boiling Point 102°C to 103°C (720 mmHg, lit.)
Molecular Formula C5H13NO2
Refractive Index n20/D 1.396
Linear Formula (CH3)2NCH(OCH3)2
MDL Number MFCD00008482
Quantity 500 mL
Synonym 1,1-Dimethoxy-N,N-dimethylmethylamine; 1,1-Dimethoxytrimethylamine
Molecular Weight (g/mol) 119.16 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.