accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4637-56-3

HONNOH(E)

CAS RN 4637-56-3

IUPAC Name: (4E)-4-(hydroxyimino)-1,4-dihydroquinolin-1-ol
Synonyms: (4E)-4-(hydroxyimino)quinolin-1-ol
Molecular Weight (g/mol): 176.18
Molecular Formula: C9H8N2O2
InChi Key: KCGKBOFRVKLKOO-CSKARUKUSA-N
SMILES: O\N=C1/C=CN(O)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results