accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 464-07-3

H3COHCH3CH3H3C

CAS RN 464-07-3

IUPAC Name: 3,3-dimethylbutan-2-ol
Synonyms: 3,3-dimethyl-2-butanol pinacolyl alcohol
Molecular Weight (g/mol): 102.18
Molecular Formula: C6H14O
InChi Key: DFOXKPDFWGNLJU-UHFFFAOYNA-N
SMILES: CC(O)C(C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)