accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3H3CH3CO(R)H3C(S)OHH3CH3CCH3H3C(R)HO(E)(E)(E)(E)(E)(E)(E)(E)(E)

CAS RN 465-42-9

IUPAC Name: (2E,4E,6E,8E,10E,12E,14E,16E,18E)-1-[(1R,4S)-4-hydroxy-1,2,2-trimethylcyclopentyl]-19-[(4R)-4-hydroxy-2,6,6-trimethylcyclohex-1-en-1-yl]-4,8,13,17-tetramethylnonadeca-2,4,6,8,10,12,14,16,18-nonaen-1-one
Synonyms: paprika extract capsanthin/capsorubin capsanthin (substance) capsanthin
Molecular Weight (g/mol): 584.89
Molecular Formula: C40H56O3
InChi Key: VYIRVAXUEZSDNC-RDJLEWNRSA-N
SMILES: C\C(\C=C\C=C(/C)\C=C\C(=O)[C@]1(C)C[C@@H](O)CC1(C)C)=C/C=C/C=C(\C)/C=C/C=C(\C)/C=C/C1=C(C)C[C@@H](O)CC1(C)C
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results