accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNSOOCl

CAS RN 465514-07-2

IUPAC Name: 3-(2-methylpyrimidin-4-yl)benzene-1-sulfonyl chloride
Synonyms: 3-(2-methylpyrimidin-4-yl)benzenesulfonyl chloride
Molecular Weight (g/mol): 268.72
Molecular Formula: C11H9ClN2O2S
InChi Key: ZLLPUDLQNCNHFM-UHFFFAOYSA-N
SMILES: CC1=NC=CC(=N1)C1=CC(=CC=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results