accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 465515-36-0

SHNNH2H2OHCl

CAS RN 465515-36-0

IUPAC Name: 1-benzothiophene-3-carboximidamide hydrate hydrochloride
Synonyms: 1-benzothiophene-3-carboximidamide hydrate hydrochloride
Molecular Weight (g/mol): 230.71
Molecular Formula: C9H11ClN2OS
InChi Key: DCDNRDYUSJNDGA-UHFFFAOYSA-N
SMILES: O.Cl.NC(=N)C1=CSC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results