accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4672-25-7

IUPAC Name: (5-phosphonopentyl)phosphonic acid
Synonyms: 5-phosphonopentylphosphonic acid
Molecular Weight (g/mol): 232.11
Molecular Formula: C5H14O6P2
InChi Key: QRKINCQKOGXVEO-UHFFFAOYSA-N
SMILES: OP(O)(=O)CCCCCP(O)(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results