accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 46745-66-8

IUPAC Name: 1-ethenyl-4-octylbenzene
Synonyms: 1-ethenyl-4-octylbenzene
Molecular Weight (g/mol): 216.37
Molecular Formula: C16H24
InChi Key: HLRQDIVVLOCZPH-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=CC=C(C=C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results